Please login to your online account to display your discounted pricing

D-Glucuronic acid, 98%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC309881000

 View more versions of this product

Catalog No. AC309881000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 194.14
Quantity 100g
Color White to off-white
EINECS Number 229-486-4
TSCA TSCA
Infrared Spectrum Authentic
Specific Rotation After 4 hrs.: +35.5° to +37.5° (20°C, 589nm) (c=6, H2O)
Water 1% max. (K.F.)
Assay Percent Range 98%
Molecular Formula C6H10O7
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Packaging Plastic bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material D-Glucuronic acid
Name Note 98%
MDL Number MFCD00064368
CAS 6556-12-3

Specifications

Chemical Name or Material D-Glucuronic acid
Name Note 98%
CAS 6556-12-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H10O7
Formula Weight 194.14
MDL Number MFCD00064368
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents