missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
α-D-Glucose Pentaacetate, MP Biomedicals™
Manufacturer: Mp Biomedicals 0210047025
Specifications
α-D-Glucose Pentaacetate | |
604-68-2 | |
MFCD00064071 | |
LPTITAGPBXDDGR-LJIZCISZSA-N | |
[(2R,3R,4S,5R,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate | |
2723636 | |
Solid |
25g | |
C16H22O11 | |
1,2,3,4,6-penta-o-acetyl-alpha-d-glucopyranose, 2r,3r,4s,5r,6r-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, a-d-glucopyranose pentaacetate, a-d-glucose pentaacetate, alpha-d-glucopyranose pentaacetate, alpha-d-glucose pentaacetate, glucopyranose, pentaacetate, .alpha.-d, penta-o-acetyl-alpha-d-glucopyranose, pentaacetyl-alpha-d-glucose, unii-1cs424ns93 | |
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C | |
390.341 | |
390.3 |
Chemical Identifiers
604-68-2 | |
390.341 | |
LPTITAGPBXDDGR-LJIZCISZSA-N | |
2723636 | |
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C |
C16H22O11 | |
MFCD00064071 | |
1,2,3,4,6-penta-o-acetyl-alpha-d-glucopyranose, 2r,3r,4s,5r,6r-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, a-d-glucopyranose pentaacetate, a-d-glucose pentaacetate, alpha-d-glucopyranose pentaacetate, alpha-d-glucose pentaacetate, glucopyranose, pentaacetate, .alpha.-d, penta-o-acetyl-alpha-d-glucopyranose, pentaacetyl-alpha-d-glucose, unii-1cs424ns93 | |
[(2R,3R,4S,5R,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate |
Safety and Handling
EINECSNumber : 210-073-2
Recommended Storage : Store at room temperature.