missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α-D-Glucose Pentaacetate, MP Biomedicals™

Manufacturer:  Mp Biomedicals 10047080

 View more versions of this product

Catalog No. ICN10047080



Specifications

Specifications

α-D-Glucose Pentaacetate
604-68-2
MFCD00064071
LPTITAGPBXDDGR-LJIZCISZSA-N
[(2R,3R,4S,5R,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
2723636
Solid
100g
C16H22O11
1,2,3,4,6-penta-o-acetyl-alpha-d-glucopyranose, 2r,3r,4s,5r,6r-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, a-d-glucopyranose pentaacetate, a-d-glucose pentaacetate, alpha-d-glucopyranose pentaacetate, alpha-d-glucose pentaacetate, glucopyranose, pentaacetate, .alpha.-d, penta-o-acetyl-alpha-d-glucopyranose, pentaacetyl-alpha-d-glucose, unii-1cs424ns93
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
390.341
390.3

Chemical Identifiers

604-68-2
390.341
LPTITAGPBXDDGR-LJIZCISZSA-N
2723636
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
C16H22O11
MFCD00064071
1,2,3,4,6-penta-o-acetyl-alpha-d-glucopyranose, 2r,3r,4s,5r,6r-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, a-d-glucopyranose pentaacetate, a-d-glucose pentaacetate, alpha-d-glucopyranose pentaacetate, alpha-d-glucose pentaacetate, glucopyranose, pentaacetate, .alpha.-d, penta-o-acetyl-alpha-d-glucopyranose, pentaacetyl-alpha-d-glucose, unii-1cs424ns93
[(2R,3R,4S,5R,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
Safety and Handling

Safety and Handling

EINECSNumber : 210-073-2

Recommended Storage : Store at room temperature.

Documents