Please login to your online account to display your discounted pricing

β-D-Glucose pentaacetate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  302960050

 View more versions of this product

Catalog No. AC302960050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation +4° to +6° (20°C, 589nm) (c=1, CHCl3)
Sulfated Ash 0.5% max.
Water 0.5% max. (K.F.)
Specific Rotation Condition + 5.00 (20.00°C c=1,CHCL3)
Color White to off-white
Quantity 5g
Chemical Name or Material β-D-Glucose pentaacetate
CAS 604-69-3
CAS Min % 97.5
CAS Max % 100.0
Physical Form White to off-white crystalline powder
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C16H22O11
Formula Weight 390.33g/mol
MDL Number MFCD00006597
Synonym 1,2,3,4,6-Penta-O-acetyl-β-D-glucose

Specifications

Chemical Name or Material β-D-Glucose pentaacetate
CAS 604-69-3
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C16H22O11
Formula Weight 390.33g/mol
MDL Number MFCD00006597
Synonym 1,2,3,4,6-Penta-O-acetyl-β-D-glucose