missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D(+)-Glucose monohydrate, 99+%, extra pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC450740250

 View more versions of this product

Catalog No. AC450740250



Description

Description

Specifications

Specifications

D(+)-Glucose monohydrate
0.1% max
14431-43-7
C6H12O6·H2O
198.17
99+%
Fibre drum
25kg
99% min. (ELSD) (HPLC)
Solubility in water: soluble.
198.17
Extra Pure

Chemical Identifiers

14431-43-7
C6H12O6·H2O
198.17
SDS Documents
Promotions

Promotions