missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-(+)-Glucose monohydrate, 99%

Manufacturer:  Alfa Aesar™ 4726336

 View more versions of this product

Catalog No. AA4726336



Specifications

Specifications

D-(+)-Glucose monohydrate
~83°C
14431-43-7
3694181
Powder
White
500g
MFCD00149450
198.18 (180.16 Anhydrous)
99%

Chemical Identifiers

14431-43-7
MFCD00149450
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents