Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Glucose diethyl mercaptal, 98%

$100.50 - $100.50

Chemical Identifiers

Chemical Name or Material D-Glucose diethyl mercaptal
CAS 1941-52-2
Molecular Formula C10H22O5S2
Formula Weight 286.41
MDL Number MFCD00004706
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2107303

  SDS
alfa aesar™
B2107303
1g Each for $100.50
Description & Specifications

Specifications

Formula Weight 286.41
Assay 98%
Quantity 1g
MDL Number MFCD00004706
Recommended Storage 2° to 8°C
CAS 1941-52-2
Melting Point 125° to 128°C
Molecular Formula C10H22O5S2
Chemical Name or Material D-Glucose diethyl mercaptal
Safety and Handling

2° to 8°C