missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Glucose ACS AR Granular, Macron Fine Chemicals™
macron
$849.26 - $849.26
Chemical Identifiers
CAS | 50-99-7 |
---|---|
Molecular Formula | C6H12O6 |
Molecular Weight (g/mol) | 180.156 |
InChI Key | GZCGUPFRVQAUEE-SLPGGIOYSA-N |
Synonym | 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form |
PubChem CID | 107526 |
ChEBI | CHEBI:42758 |
IUPAC Name | (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
SMILES | C(C(C(C(C(C=O)O)O)O)O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
MK4912500
|
Avantor
491212 |
500 g | POLYSTORMOR |
Case of 4 Each for $849.26
|
|
||||
Chemical Identifiers
50-99-7 | |
180.156 | |
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
CHEBI:42758 | |
C(C(C(C(C(C=O)O)O)O)O)O |
C6H12O6 | |
GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
107526 | |
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
Specifications
50-99-7 | |
C6H12O6 | |
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
C(C(C(C(C(C=O)O)O)O)O)O | |
180.156 | |
CHEBI:42758 | |
ACS | |
D-Glucose, Anhydrous |
POLYSTORMOR | |
500 g | |
GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal | |
107526 | |
180.16 | |
Granules |