Please login to your online account to display your discounted pricing

α-D-Glucose-1-phosphate dipotassium salt dihydrate, 98.8%, MP Biomedicals™

$58.99 - $139.23

Chemical Identifiers

Chemical Name or Material α-D-Glucose-1-phosphate dipotassium salt dihydrate
CAS 5996-14-5
Physical Form Powder
Assay Percent Range 98.80%
Molecular Formula C6H11O9PK2·2H2O
Formula Weight 372.3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19467380

 
mp biomedicals
19467380
100mg Each for $139.23

ICN19467325

 
mp biomedicals
19467325
25mg Each for $58.99
Description & Specifications

Specifications

Formula Weight 372.3
Physical Form Powder
Color White
Quantity 100mg
Chemical Name or Material α-D-Glucose-1-phosphate dipotassium salt dihydrate
CAS 5996-14-5
Molecular Formula C6H11O9PK2·2H2O
Assay Percent Range 98.80%