Please login to your online account to display your discounted pricing

α-D-Glucose-1-phosphate dipotassium salt dihydrate, 98.8%, MP Biomedicals™


Quantity 1g

Manufacturer: mp biomedicals  0219467391

 View more versions of this product

Catalog No. ICN19467391

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1g
Chemical Name or Material α-D-Glucose-1-phosphate dipotassium salt dihydrate
CAS 5996-14-5
Physical Form White powder
Assay Percent Range 98.80%
Molecular Formula C6H11O9PK2·2H2O
Formula Weight 372.3g/mol

Specifications

Chemical Name or Material α-D-Glucose-1-phosphate dipotassium salt dihydrate
CAS 5996-14-5
Physical Form White powder
Assay Percent Range 98.80%
Molecular Formula C6H11O9PK2·2H2O
Formula Weight 372.3g/mol