Please login to your online account to display your discounted pricing

D-Gluconic acid, zinc salt hydrate, 97+%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  392015000

 View more versions of this product

Catalog No. AC392015000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Plastic bottle
Water 11.6% max.
Color Light beige
Quantity 500g
Chemical Name or Material D-Gluconic acid, zinc salt hydrate
Physical Form Light beige granular powder
Assay 97.0% min. (on dry substance)
Assay Percent Range 97+%
Molecular Formula C12H22O14Zn·xH2O
Linear Formula [CH2(OH)(CH(OH))4COO]2Zn·xH2O
Formula Weight 455.68g/mol
MDL Number MFCD04037230
Synonym Zinc gluconate hydrate

Specifications

Chemical Name or Material D-Gluconic acid, zinc salt hydrate
Assay 97.0% min. (on dry substance)
Assay Percent Range 97+%
Molecular Formula C12H22O14Zn·xH2O
Linear Formula [CH2(OH)(CH(OH))4COO]2Zn·xH2O
MDL Number MFCD04037230
Synonym Zinc gluconate hydrate