Please login to your online account to display your discounted pricing

D-Gluconic acid, zinc salt hydrate, 97+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material D-Gluconic acid, zinc salt hydrate
Assay 97.0% min. (on dry substance)
Assay Percent Range 97+%
Molecular Formula C12H22O14Zn·xH2O
Linear Formula [CH2(OH)(CH(OH))4COO]2Zn·xH2O
MDL Number MFCD04037230
Synonym Zinc gluconate hydrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC392015000

  SDS
thermo fisher scientific
392015000
Plastic bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Plastic bottle
Water 11.6% max.
Color Light beige
Quantity 500g
Chemical Name or Material D-Gluconic acid, zinc salt hydrate
Physical Form Light beige granular powder
Assay 97.0% min. (on dry substance)
Assay Percent Range 97+%
Molecular Formula C12H22O14Zn·xH2O
Linear Formula [CH2(OH)(CH(OH))4COO]2Zn·xH2O
Formula Weight 455.68g/mol
MDL Number MFCD04037230
Synonym Zinc gluconate hydrate