Learn More
D-Gluconic acid, 50% aq. soln., Thermo Scientific Chemicals
$51.37 - $137.51
Chemical Identifiers
CAS | 526-95-4 |
---|---|
Molecular Formula | C6H12O7 |
Molecular Weight (g/mol) | 196.155 |
MDL Number | MFCD00004240 |
InChI Key | RGHNJXZEOKUKBD-SQOUGZDYSA-N |
Synonym | gluconic acid, d-gluconic acid, dextronic acid, maltonic acid, glycogenic acid, glosanto, gluconate, pentahydroxycaproic acid, gluconic acid, d, d-gluconate |
PubChem CID | 10690 |
ChEBI | CHEBI:33198 |
IUPAC Name | (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid |
SMILES | C(C(C(C(C(C(=O)O)O)O)O)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA16806AP
|
Thermo Scientific Chemicals
A16806AP |
500 mL |
Each for $51.37
|
|
|||||
AAA168060F
|
Thermo Scientific Chemicals
A168060F |
2500 mL |
Each for $137.51
|
|
|||||
Description
Gluconic acid occurs naturally in fruit, honey, and wine. As a food additive it is an acidity regulator. It is also used in cleaning products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGluconic acid occurs naturally in fruit, honey, and wine. As a food additive it is an acidity regulator. It is also used in cleaning products.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
526-95-4 | |
196.155 | |
RGHNJXZEOKUKBD-SQOUGZDYSA-N | |
10690 | |
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid |
C6H12O7 | |
MFCD00004240 | |
gluconic acid, d-gluconic acid, dextronic acid, maltonic acid, glycogenic acid, glosanto, gluconate, pentahydroxycaproic acid, gluconic acid, d, d-gluconate | |
CHEBI:33198 | |
C(C(C(C(C(C(=O)O)O)O)O)O)O |
Specifications
526-95-4 | |
MFCD00004240 | |
1726055 | |
gluconic acid, d-gluconic acid, dextronic acid, maltonic acid, glycogenic acid, glosanto, gluconate, pentahydroxycaproic acid, gluconic acid, d, d-gluconate | |
RGHNJXZEOKUKBD-SQOUGZDYSA-N | |
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid | |
10690 | |
196.16 (Anhydrous) | |
50% aq. soln. | |
+10° (Neat) | |
D-Gluconic acid |
C6H12O7 | |
UN3265 | |
144456 | |
Soluble in water. | |
C(C(C(C(C(C(=O)O)O)O)O)O)O | |
196.155 | |
CHEBI:33198 | |
1.234 | |
500 mL | |
1.418 |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 208-401-4
RTECSNumber : LZ5057100
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only