missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D(+)-Galactose, 99+%, Acros Organics™

Manufacturer:  Acros Organics 150610010

 View more versions of this product

Catalog No. AC150610010



Specifications

Specifications

(10% aq. soln.) clear, colorless
6.2
+ 79.75 (20.00°C c=10,H2O,NH3)
0.1% max. (on dry substance)
0.3% max. (K.F.)
165.0°C to 168.0°C
59-23-4
C6H12O6
31, 295
180.16
≥99%
D(+)-Galactose
Authentic
+ 79.75
Negligible
White
1kg
99+%
MFCD00063833
15, 4365
Powder

Chemical Identifiers

59-23-4
C6H12O6
MFCD00063833
SDS Documents