Please login to your online account to display your discounted pricing

Alfa Aesar™ D-(+)-Galactose, 98%

$31.55 - $84.51

Chemical Identifiers

Chemical Name or Material D-(+)-Galactose
CAS 59-23-4
Molecular Formula C6H12O6
Formula Weight 180.16
MDL Number MFCD00151230
Beilstein 1724619
Merck Index 14,4335
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA128130B

 
alfa aesar™
A128130B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1281330

 
alfa aesar™
A1281330
250g Each for $84.51

AAA1281318

 
alfa aesar™
A1281318
50g Each for $31.55
Description & Specifications

Specifications

Formula Weight 180.16
Chemical Name or Material D-(+)-Galactose
TSCA TSCA
Assay 98%
Molecular Formula C6H12O6
MDL Number MFCD00151230
CAS 59-23-4
Beilstein 1724619
Merck Index 14,4335
Melting Point 164° to 168°C
RTECS Number LW5490000
Optical Rotation +80° (c=5 in Water, 24h)
Quantity 1000g

Applications
Galactose is a simple monosaccharide used as a component of the galactosyltransferase labeling buffer. It serves as an energy source and an important constituent of glycolipids and glycoproteins. Since it is a component of antigens, it plays a vital role in the determination of blood type within the ABO blood group system.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.