Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Galactosamine hydrochloride, 98%

$36.32 - $120.69

Chemical Identifiers

Chemical Name or Material D-Galactosamine hydrochloride
CAS 1772-03-8
Molecular Formula C6H13NO5·HCl
Formula Weight 215.64
MDL Number MFCD00135830
Beilstein 3697825
Merck Index 14,4334
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0746203

 
alfa aesar™
L0746203
1g Each for $120.69

AAL07462MD

 
alfa aesar™
L07462MD
250mg Each for $36.32
Description & Specifications

Specifications

Formula Weight 215.64
Chemical Name or Material D-Galactosamine hydrochloride
TSCA TSCA
Assay 98%
Molecular Formula C6H13NO5·HCl
MDL Number MFCD00135830
Beilstein 3697825
Melting Point ≈183°C
Quantity 1g
CAS 1772-03-8
Merck Index 14,4334
RTECS Number LW5500000
Optical Rotation +97° (c=1 in Water)