D(+)-Fucose, 98+%, Thermo Scientific™
D (+)-Fucose, 98+%, CAS # 3615-37-0, also known as aldehyde-D-fucose, is an enantiomer of an aldehyde-L-fucose found in plants. It shows several biological activities, including cellular adhesion.
Supplier:  Thermo Scientific Chemicals 225632500
Catalog No. AC225632500
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• D (+)-Fucose can be found mainly in plants and shows several biological activities, including cellular adhesion
Applications
• D-Fucose is usually found in plants and some microorganisms, as a relatively simple glycoside comprising only a few sugar units
• It can be used as a sole carbon source in some microorganisms
• It may be used to study enzymes involved in its mutarotation to form L-fucose
• This compound is used as a reference compound in the analysis of carbohydrate metabolites
SDS Documents