Please login to your online account to display your discounted pricing

D(+)-Fucose, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  225630010

 View more versions of this product

Catalog No. AC225630010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 164.16
Assay ≥98.5%
Quantity 1g
Color White
Beilstein 31, 76
Merck Index 15, 4307
Specific Rotation Condition + 76.00 (20.00°C c=4,H2O,NH3)
Packaging Glass bottle
Appearance of Solution (5% aq. soln.) clear, colorless
Assay Percent Range 99%
Molecular Formula C6H12O5
Chemical Name or Material D(+)-Fucose
MDL Number MFCD00135603
Specific Rotation +74° to +78° (20°C, 589nm) (C=4, H2O, NH3)
Water 0.5% max. (K.F.)
Synonym Rhodeose
Infrared Spectrum Authentic
CAS 3615-37-0

Specifications

Chemical Name or Material D(+)-Fucose
CAS 3615-37-0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H12O5
Formula Weight 164.16
MDL Number MFCD00135603
Synonym Rhodeose