Please login to your online account to display your discounted pricing

Alfa Aesar™ D-(+)-Fucose, 99%

$105.78 - $426.42

Chemical Identifiers

Chemical Name or Material D-(+)-Fucose
CAS 3615-37-0
Molecular Formula C6H12O5
Formula Weight 164.16
MDL Number MFCD00135603
Beilstein 1723320
Merck Index 14,4278
Synonym 6-Deoxy-D-galactose; Rhodeose
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1823403

  SDS
alfa aesar™
A1823403
1g Each for $105.78

AAA1823406

  SDS
alfa aesar™
A1823406
5g Each for $426.42
Description & Specifications

Specifications

Formula Weight 164.16
Chemical Name or Material D-(+)-Fucose
TSCA TSCA
Assay 99%
Molecular Formula C6H12O5
MDL Number MFCD00135603
CAS 3615-37-0
Beilstein 1723320
Merck Index 14,4278
Synonym 6-Deoxy-D-galactose; Rhodeose
Melting Point 135° to 141°C
Optical Rotation +76° (c=10 in Water)
Quantity 1g