Please login to your online account to display your discounted pricing

D-Fructose (Crystalline/USP), Fisher Chemical


Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLL96212

 View more versions of this product

Catalog No. L96-212

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 180.16
pH 5 to 7
Melting Point 103°C
Quantity 2.5kg
Purity Grade USP
Assay 98.0 to 102.0% (USP)
Color White
Alpha Vector Fructose
Physical Form Solid
Molecular Formula C6H12O6
Calcium and Magnesium 0.005% max. (USP)
Heavy Metals (as Pb) 5ppm max. (USP)
Loss on Drying 0.5% max. (USP)
Synonym β-D-Fructose
Hydroxymethyl Furfural Pass Test
Packaging Poly Bottle
Identification Pass Test (USP)
Arsenic (As) 1ppm max. (USP)
Sulfate 0.025% max. (USP)
Residual Solvents Meets Requirements (USP)
Chloride 0.018% max. (USP)
Acidity Pass Test (USP)
Ignition Residue 0.5% max. (USP)
MDL Number MFCD00148910
ChemAlert Storage Symbol Gray
Chemical Name or Material D-Fructose
CAS 57-48-7

Specifications

Chemical Name or Material D-Fructose
CAS 57-48-7
Purity Grade USP
Physical Form Solid
Assay 98.0 to 102.0% (USP)
Molecular Formula C6H12O6
Formula Weight 180.16
MDL Number MFCD00148910
Synonym β-D-Fructose
Accessories
 

SDS & Safety Documents