missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Fructose (Crystalline/USP), Fisher Chemical

Manufacturer:  Thermo Scientific FLL9612

 View more versions of this product

Catalog No. L96-12



Specifications

Specifications

Pass Test (USP)
D-Fructose
0.5% max. (USP)
Poly Pail
White
12kg
57-48-7
MFCD00148910
BJHIKXHVCXFQLS-UYFOZJQFSA-N
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
5984
180.16
98.0 to 102.0% (USP)
Fructose
Pass Test (USP)
0.5% max. (USP)
Meets Requirements (USP)
103°C
5 to 7
C6H12O6
d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
C(C(C(C(C(=O)CO)O)O)O)O
180.156
CHEBI:48095
Solid
USP

Chemical Identifiers

57-48-7
180.156
BJHIKXHVCXFQLS-UYFOZJQFSA-N
5984
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
C6H12O6
MFCD00148910
d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
CHEBI:48095
C(C(C(C(C(=O)CO)O)O)O)O
Safety and Handling

Safety and Handling

SDS Documents