Please login to your online account to display your discounted pricing

D-Fructose (Crystalline/USP), Fisher Chemical


Quantity 100 lb.
Packaging Fiber Drum

Manufacturer: fisher scientific company  FLL96100LB

 View more versions of this product

Catalog No. L96-100LB

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 180.16
Melting Point 103°C
pH 5 to 7
Quantity 100 lb.
Purity Grade USP
Physical Form Solid
Assay 98.0 to 102.0% (USP)
Molecular Formula C6H12O6
Alpha Vector Fructose
Color White
Residual Solvents Meets Requirements (USP)
Acidity Pass Test (USP)
Heavy Metals (as Pb) 5ppm max. (USP)
Identification Pass Test (USP)
Ignition Residue 0.5% max. (USP)
Loss on Drying 0.5% max. (USP)
Packaging Fiber Drum
MDL Number MFCD00148910
Synonym β-D-Fructose
Arsenic (As) 1ppm max. (USP)
Chemical Name or Material D-Fructose
Hydroxymethyl Furfural Pass Test
CAS 57-48-7
ChemAlert Storage Symbol Gray
Sulfate 0.025% max. (USP)
Chloride 0.018% max. (USP)
Calcium and Magnesium 0.005% max. (USP)

Specifications

Chemical Name or Material D-Fructose
CAS 57-48-7
Purity Grade USP
Physical Form Solid
Assay 98.0 to 102.0% (USP)
Molecular Formula C6H12O6
Formula Weight 180.16
MDL Number MFCD00148910
Synonym β-D-Fructose
Accessories
 

SDS & Safety Documents