Please login to your online account to display your discounted pricing

D-(-)-Fructose, BAKER ANALYZED™ Biochemical Reagent, J.T.Baker™

High quality biochemicals for research use

$99.36 - $99.36

Chemical Identifiers

Chemical Name or Material D-(-)-Fructose
CAS 57-48-7
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Molecular Formula C6H12O6
Formula Weight 180.16g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002680

 
avantor performance
M556-05
Poly Bottle 100g Each for $99.36
Description & Specifications

Specifications

Packaging Poly Bottle
Quantity 100g
Chemical Name or Material D-(-)-Fructose
CAS 57-48-7
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Molecular Formula C6H12O6
Formula Weight 180.16g/mol