missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-(-)-Fructose, BAKER ANALYZED™ Biochemical Reagent, J.T.Baker™

High quality biochemicals for research use

Manufacturer:  Avantor J.T.Baker M55606

 View more versions of this product

Catalog No. 02002682



Description

Description

Specifications

Specifications

D-(-)-Fructose
5kg
C6H12O6
BJHIKXHVCXFQLS-UYFOZJQFSA-N
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
5984
180.16
Poly Pail
57-48-7
3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, arabino-hexulose, d---fructose, d--fructose, d-levulose, fructose, d, furucton, keto-d-fructose, krystar 300, sugar, fruit
C(C(C(C(C(=O)CO)O)O)O)O
180.156
CHEBI:48095
BAKER ANALYZED™ Biochemical Reagent

Chemical Identifiers

57-48-7
180.156
3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, arabino-hexulose, d---fructose, d--fructose, d-levulose, fructose, d, furucton, keto-d-fructose, krystar 300, sugar, fruit
CHEBI:48095
C(C(C(C(C(=O)CO)O)O)O)O
C6H12O6
BJHIKXHVCXFQLS-UYFOZJQFSA-N
5984
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SDS
Documents
Product Certifications
Promotions

Promotions