Please login to your online account to display your discounted pricing

D(+)-Fenchone, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC366290250

 View more versions of this product

Catalog No. AC366290250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 152.24
Physical Form Liquid
Quantity 25g
Color Clear colorless to pale yellow
Beilstein 07, IV, 212
Merck Index 15, 4000
Specific Rotation Condition +59.00 (20.00°C neat)
Density 0.9500g/mL
Refractive Index 1.4610 to 1.4640 (20°C, 589nm)
Name Note 97%
CAS 4695-62-9
MDL Number MFCD00070689
Packaging Glass bottle
Infrared Spectrum Authentic
Specific Rotation +55° to +63° (20°C, 589nm) (neat)
Synonym D(+)-1, 3, 3-Trimethylbicylo[2.2.1]heptan-2-one
Molecular Formula C10H16O
Assay Percent Range 97%
Chemical Name or Material D(+)-Fenchone

Specifications

Chemical Name or Material D(+)-Fenchone
Name Note 97%
CAS 4695-62-9
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H16O
Formula Weight 152.24
MDL Number MFCD00070689
Synonym D(+)-1, 3, 3-Trimethylbicylo[2.2.1]heptan-2-one