Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Erythrose, syrup, ca 70% w/v, >75% dry wt. basis

$52.88 - $128.69

Chemical Identifiers

Chemical Name or Material D-Erythrose
CAS 583-50-6
Name Note syrup
Molecular Formula C4H8O4
Formula Weight 120.1
MDL Number MFCD00006970
Beilstein 1721698
Merck Index 14,3690
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2109303

  SDS
alfa aesar™
B2109303
1g Each for $128.69

AAB2109301

  SDS
alfa aesar™
B2109301
0.25g Each for $52.88
Description & Specifications

Specifications

Formula Weight 120.1
Chemical Name or Material D-Erythrose
Molecular Formula C4H8O4
MDL Number MFCD00006970
CAS 583-50-6
Quantity 1g
Concentration ≈70% w/v
Beilstein 1721698
Recommended Storage 2° to 8°C
Optical Rotation −33° (c=2 in Water, 72H)
Assay Percent Notes dry wt. basis
Name Note syrup
Merck Index 14,3690
Assay >75%

Applications
Erythrose is an anti-cancer agent. In vitro, ~4 mM erythrose effectively kills cancer cells for tested cell lines (in vitro), such as: lung, brain, breast and colorectal cancers. D-Erythrose inhibits tumor growth in mice (in vivo). It is used to study the mechanisms of organic micro spherule formation and Maillard (glycation) reactions.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Safety and Handling

2° to 8°C