missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-erythro-Sphingosine, 99+%

$156.64 - $277.99

Chemical Identifiers

CAS 123-78-4
Molecular Formula C18H37NO2
Molecular Weight (g/mol) 299.499
MDL Number MFCD00036751
InChI Key WWUZIQQURGPMPG-KRWOKUGFSA-N
Synonym 2s,3r,4e-2-amino-4-octadecene-1,3-diol, 4-sphingenine, 4-trans-sphingenine, cerebroside, d-erythro-sphingosine, d-sphingosine, sphing-4-enine, sphingenine, sphingosine, trans-d-erythro-2-amino-4-octadecene-1,3-diol
PubChem CID 5280335
ChEBI CHEBI:16393
IUPAC Name (E,2S,3R)-2-aminooctadec-4-ene-1,3-diol
SMILES CCCCCCCCCCCCCC=CC(C(CO)N)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63584MA View Documents Alfa Aesar™
J63584MA
10mg Each for $156.64
 
AAJ63584MB View Documents Alfa Aesar™
J63584MB
25mg Each for $277.99
 
Description

Description

Applications
D-erythro-Sphingosine is used to inhibits protein kinase C and calmodulin-dependent enzymes, induces apoptosis. And also used to constituent of cell membranes.

Solubility
Soluble in ethanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

123-78-4
299.499
WWUZIQQURGPMPG-KRWOKUGFSA-N
5280335
(E,2S,3R)-2-aminooctadec-4-ene-1,3-diol
C18H37NO2
MFCD00036751
2s,3r,4e-2-amino-4-octadecene-1,3-diol, 4-sphingenine, 4-trans-sphingenine, cerebroside, d-erythro-sphingosine, d-sphingosine, sphing-4-enine, sphingenine, sphingosine, trans-d-erythro-2-amino-4-octadecene-1,3-diol
CHEBI:16393
CCCCCCCCCCCCCC=CC(C(CO)N)O

Specifications

White
123-78-4
MFCD00036751
14,8747
Soluble in ethanol.
CCCCCCCCCCCCCC=CC(C(CO)N)O
299.499
CHEBI:16393
Powder
D-erythro-Sphingosine
10mg
C18H37NO2
1727294
2s,3r,4e-2-amino-4-octadecene-1,3-diol, 4-sphingenine, 4-trans-sphingenine, cerebroside, d-erythro-sphingosine, d-sphingosine, sphing-4-enine, sphingenine, sphingosine, trans-d-erythro-2-amino-4-octadecene-1,3-diol
WWUZIQQURGPMPG-KRWOKUGFSA-N
(E,2S,3R)-2-aminooctadec-4-ene-1,3-diol
5280335
299.5
≥99%
Safety and Handling

Safety and Handling

EINECSNumber : 204-651-3

TSCA : No

Recommended Storage : Store at -20°C