Please login to your online account to display your discounted pricing

D-Cystine 98%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  133982500

 View more versions of this product

Catalog No. AC133982500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 240.29
Physical Form Fine Crystalline Powder
Quantity 250mg
Color White
Merck Index 15, 2779
EINECS Number 206-486-2
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Appearance of Solution (1% soln. in 1N HCl) clear, colorless
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula [-SCH2CH(NH2)CO2H]2
Molecular Formula C6H12N2O4S2
Assay Percent Range 98%
Name Note 98%
Chemical Name or Material D-Cystine
MDL Number MFCD00002610
CAS 349-46-2
Specific Rotation +211° to +217° (20°C, 589nm) (c=1, 1N HCl)
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym D(+)-3, 3';-Dithiobis(2-aminopropanoic acid)
Sulfated Ash 0.1% max.

Specifications

Chemical Name or Material D-Cystine
Name Note 98%
CAS 349-46-2
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H12N2O4S2
Linear Formula [-SCH2CH(NH2)CO2H]2
Formula Weight 240.29
MDL Number MFCD00002610
Synonym D(+)-3, 3';-Dithiobis(2-aminopropanoic acid)
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents