Please login to your online account to display your discounted pricing

Alfa Aesar™ D-(+)-Cellotriose, 95%

Chemical Identifiers

Chemical Name or Material D-(+)-Cellotriose
CAS 33404-34-1
Formula Weight 504.44
MDL Number MFCD00069842
Beilstein 100353
Synonym Cellotriose; 1,4-β-D-Cellotriose
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67702LB0

  SDS
alfa aesar™
J67702LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 504.44
Chemical Name or Material D-(+)-Cellotriose
CAS 33404-34-1
Beilstein 100353
Storage Note 1 -30° to -10°C
Quantity 5mg
Assay 95%
MDL Number MFCD00069842
Sensitivity Hygroscopic
Synonym Cellotriose; 1,4-β-D-Cellotriose
Solubility Information Soluble in water at 50mg/mL
Color White to off-white
Physical Form Powder
Safety and Handling

-30° to -10°C