Please login to your online account to display your discounted pricing

Alfa Aesar™ D-(+)-Cellotetraose, 95%

Chemical Identifiers

Chemical Name or Material D-(+)-Cellotetraose
CAS 38819-01-1
Formula Weight 666.58
MDL Number MFCD00079079
Beilstein 78869
Synonym Cellotetraose
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67583LB0

 
alfa aesar™
J67583LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 666.58
Chemical Name or Material D-(+)-Cellotetraose
MDL Number MFCD00079079
Physical Form Powder
Assay 95%
CAS 38819-01-1
Beilstein 78869
Synonym Cellotetraose
Storage Note 1 -30° to -10°C
Sensitivity Hygroscopic
Color White to off-white
Quantity 5mg
Solubility Information Soluble at 25mg/mL in water

  • Derived from cellulose degradation
  • Used by cellulosic bacteria as an energy source
  • Useful for characterization and identification of oligosaccharide metabolizing enzymes such as endoglucanases and _x001A_-glucosidases

Safety and Handling

-30° to -10°C