Please login to your online account to display your discounted pricing

Alfa Aesar™ D-(+)-Celloheptaose, 95%


Quantity 2mg

Manufacturer: alfa aesar™  J678038PL

 View more versions of this product

Catalog No. AAJ678038PL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 1153.02
Chemical Name or Material D-(+)-Celloheptaose
Physical Form Powder
Assay 95%
MDL Number MFCD15145329
CAS 52646-27-2
Synonym Celloheptaose
Storage Note 1 -30° to -10°C
Sensitivity Hygroscopic
Color White to off-white
Quantity 2mg

Specifications

Chemical Name or Material D-(+)-Celloheptaose
CAS 52646-27-2
Formula Weight 1153.02
MDL Number MFCD15145329
Synonym Celloheptaose
Accessories

-30° to -10°C

SDS & Safety Documents