Please login to your online account to display your discounted pricing

Alfa Aesar™ D-(+)-Celloheptaose, 95%

Chemical Identifiers

Chemical Name or Material D-(+)-Celloheptaose
CAS 52646-27-2
Formula Weight 1153.02
MDL Number MFCD15145329
Synonym Celloheptaose
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67803LB0

  SDS
alfa aesar™
J67803LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ678038PL

  SDS
alfa aesar™
J678038PL
2mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67803MCR

  SDS
alfa aesar™
J67803MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 1153.02
Chemical Name or Material D-(+)-Celloheptaose
CAS 52646-27-2
MDL Number MFCD15145329
Synonym Celloheptaose
Assay 95%
Color White to off-white
Storage Note 1 -30° to -10°C
Quantity 5mg
Sensitivity Hygroscopic
Physical Form Powder
Safety and Handling

-30° to -10°C