Please login to your online account to display your discounted pricing

D-Camphoroquinone, ACROS Organics™

$421.95 - $421.95

Chemical Identifiers

Chemical Name or Material D-Camphoroquinone
CAS 10334-26-6
Molecular Formula C10 H14 O2
Formula Weight 166.22
MDL Number MFCD00082863
Synonym D-2, 3-Bornanedione
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC402750250

  SDS
thermo fisher scientific
402750250
Glass bottle 25g Each for $421.95
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 166.22
Quantity 25g
Color Yellow
EINECS Number 220-446-1
TSCA TSCA
Melting Point 200° to 204°C
Infrared Spectrum Authentic
Synonym D-2, 3-Bornanedione
Packaging Glass bottle
Chemical Name or Material D-Camphoroquinone
Molecular Formula C10 H14 O2
CAS 10334-26-6
Specific Rotation -66° to -74° (20°C, 589nm) (c=4, ethanol) after 24 hrs.
MDL Number MFCD00082863