missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D(+)-Camphor, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 227841000
Specifications
D(+)-Camphor | |
Authentic | |
+ 42.50 | |
White | |
100g | |
464-49-3 | |
C10H16O | |
13,61 | |
+-2-bornanone, +-bornan-2-one, +-camphor, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, 1r,4r-camphor, 1r-+-camphor, camphor d, d-camphor, r-+-camphor, r-camphor | |
DSSYKIVIOFKYAU-XCBNKYQSSA-N | |
(1R,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one | |
159055 | |
152.24 | |
97% |
64°C | |
Plastic bottle | |
204.0°C | |
176.0°C to 180.0°C | |
97% | |
96% min. (GC) | |
MFCD00064149 | |
15, 1734 | |
Solubility in water: 1g/800mL water (25°C) (giving a colloidal solutio. Other solubilities: at 25°C 1 g dissolves in 1 mL alcohol,1 mL ether,,0.5 mL chloroform,0.4 mL benzene,0.4 mL acetone,,1.5 mL turpentine oil,0.5 mL glacial acetic acid | |
CC1(C2CCC1(C(=O)C2)C)C | |
152.24 | |
CHEBI:15396 | |
Crystals |
Chemical Identifiers
464-49-3 | |
152.24 | |
DSSYKIVIOFKYAU-XCBNKYQSSA-N | |
159055 | |
(1R,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one |
C10H16O | |
MFCD00064149 | |
+-2-bornanone, +-bornan-2-one, +-camphor, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, 1r,4r-camphor, 1r-+-camphor, camphor d, d-camphor, r-+-camphor, r-camphor | |
CHEBI:15396 | |
CC1(C2CCC1(C(=O)C2)C)C |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.
Harmful if swallowed.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cauti
GHS Signal Word: Warning
EINECSNumber : 207-355-2
RTECSNumber : EX1260000
TSCA : TSCA