missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Aspartic acid dibenzyl ester p-toluenesulfonate, 95%

$83.42 - $303.80

Chemical Identifiers

CAS 4079-64-5
Molecular Formula C25H27NO7S
Molecular Weight (g/mol) 485.551
MDL Number MFCD00035102
InChI Key HLMUYZYLPUHSNV-PKLMIRHRSA-N
Synonym h-d-asp obzl-obzl.tosoh, d-aspartic acid dibenzyl ester-p-toluenesulfonate, r-dibenzyl 2-aminosuccinate 4-methylbenzenesulfonate, d-aspartic acid dibenzyl ester tosylate, d-aspartic acid dibenzyl ester p-toluenesulfonate, d-aspartic acid dibenzyl ester 4-toluenesulfonate salt, d-aspdb tsoh, 1,4-dibenzyl 2r-2-aminobutanedioate; para-toluene sulfonate, h-d-asp obzl-obzl tos, d,l-aspartic acid dibenzyl ester-p-toluenesulfonate
PubChem CID 25188997
IUPAC Name dibenzyl (2R)-2-aminobutanedioate;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CC(C(=O)OCC2=CC=CC=C2)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6547514 View Documents Alfa Aesar™
H6547514
25g Each for $83.42
 
AAH6547522 View Documents Alfa Aesar™
H6547522
100g Each for $303.80
 
Specifications

Chemical Identifiers

4079-64-5
485.551
HLMUYZYLPUHSNV-PKLMIRHRSA-N
25188997
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CC(C(=O)OCC2=CC=CC=C2)N
C25H27NO7S
MFCD00035102
h-d-asp obzl-obzl.tosoh, d-aspartic acid dibenzyl ester-p-toluenesulfonate, r-dibenzyl 2-aminosuccinate 4-methylbenzenesulfonate, d-aspartic acid dibenzyl ester tosylate, d-aspartic acid dibenzyl ester p-toluenesulfonate, d-aspartic acid dibenzyl ester 4-toluenesulfonate salt, d-aspdb tsoh, 1,4-dibenzyl 2r-2-aminobutanedioate; para-toluene sulfonate, h-d-asp obzl-obzl tos, d,l-aspartic acid dibenzyl ester-p-toluenesulfonate
dibenzyl (2R)-2-aminobutanedioate;4-methylbenzenesulfonic acid

Specifications

4079-64-5
MFCD00035102
h-d-asp obzl-obzl.tosoh, d-aspartic acid dibenzyl ester-p-toluenesulfonate, r-dibenzyl 2-aminosuccinate 4-methylbenzenesulfonate, d-aspartic acid dibenzyl ester tosylate, d-aspartic acid dibenzyl ester p-toluenesulfonate, d-aspartic acid dibenzyl ester 4-toluenesulfonate salt, d-aspdb tsoh, 1,4-dibenzyl 2r-2-aminobutanedioate; para-toluene sulfonate, h-d-asp obzl-obzl tos, d,l-aspartic acid dibenzyl ester-p-toluenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CC(C(=O)OCC2=CC=CC=C2)N
485.551
485.55
95%
25g
C25H27NO7S
6311519
HLMUYZYLPUHSNV-PKLMIRHRSA-N
dibenzyl (2R)-2-aminobutanedioate;4-methylbenzenesulfonic acid
25188997
Solid
155°C to 158°C
D-Aspartic acid dibenzyl ester p-toluenesulfonate
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C