missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Aspartic acid 1-methyl ester, 98%

Manufacturer:  Alfa Aesar™ H6365803

 View more versions of this product

Catalog No. AAH6365803



Description

Description

Applications
D-Aspartic acid 1-methyl ester is used as a intermediate for pharmaceutical and chemical research.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

D-Aspartic acid 1-methyl ester
C5H9NO4
h-d-asp-ome, r-3-amino-4-methoxy-4-oxobutanoic acid, d-asp-ome, d-aspartic acid 1-methyl ester, d-aspartic acid,1-methyl ester, 3r-3-amino-4-methoxy-4-oxobutanoic acid
COC(=O)C(CC(=O)O)N
147.13
147.13
98%
65414-78-0
MFCD03424227
SWWBMHIMADRNIK-GSVOUGTGSA-N
(3R)-3-amino-4-methoxy-4-oxobutanoic acid
7020023
Solid
1g

Chemical Identifiers

65414-78-0
147.13
SWWBMHIMADRNIK-GSVOUGTGSA-N
7020023
COC(=O)C(CC(=O)O)N
C5H9NO4
MFCD03424227
h-d-asp-ome, r-3-amino-4-methoxy-4-oxobutanoic acid, d-asp-ome, d-aspartic acid 1-methyl ester, d-aspartic acid,1-methyl ester, 3r-3-amino-4-methoxy-4-oxobutanoic acid
(3R)-3-amino-4-methoxy-4-oxobutanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents