Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
missing translation for 'pleaseLogIn'

Alfa Aesar™ D-Arginine methyl ester dihydrochloride, 98%

Manufacturer:  Alfa Aesar™ H6269503

 View more versions of this product

Catalog No. AAH6269503


 

Specifications

Specifications

D-Arginine methyl ester dihydrochloride
98%
MFCD00153417
XQYZOBNLCUAXLF-ZJIMSODOSA-N
methyl (2R)-2-amino-5-(diaminomethylideneamino)pentanoate;dihydrochloride
12714066
Solid
Hygroscopic
78851-84-0
C7H18Cl2N4O2
H-D-Arg-OMe.2HCl
COC(=O)C(CCCN=C(N)N)N.Cl.Cl
261.147
261.15
1g

Chemical Identifiers

78851-84-0
261.147
XQYZOBNLCUAXLF-ZJIMSODOSA-N
12714066
C7H18Cl2N4O2
MFCD00153417
H-D-Arg-OMe.2HCl
SDS Documents