Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Arginine methyl ester dihydrochloride, 98%

Chemical Identifiers

Chemical Name or Material D-Arginine methyl ester dihydrochloride
CAS 78851-84-0
Formula Weight 261.15
MDL Number MFCD00153417
Synonym H-D-Arg-OMe.2HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6269503

 
alfa aesar™
H6269503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6269506

 
alfa aesar™
H6269506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 261.15
Chemical Name or Material D-Arginine methyl ester dihydrochloride
Physical Form Solid
Assay 98%
MDL Number MFCD00153417
CAS 78851-84-0
Synonym H-D-Arg-OMe.2HCl
Quantity 1g
Sensitivity Hygroscopic