Please login to your online account to display your discounted pricing

Alfa Aesar™ [D-Arg&superscript_0;, Hyp&superscript_3;, Thi&superscript_5;,&superscript_8;, D-Phe&superscript_7;]-Adamantaneacetyl Bradykinin

For cellular and molecular biology applications

Specifications

Product Type [D-Arg0, Hyp3, Thi5,8, D-Phe7]-Adamantaneacetyl Bradykinin
Molecular Weight 1469.75
Storage Requirements -30° to -10°C
Sequence N-Adamantaneacetyl-D-Arg-Arg-Pro-Hyp-Gly-Thi-Ser-D-Phe-Thi-Arg
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66404LB0

 
alfa aesar™
J66404LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66404MCR

 
alfa aesar™
J66404MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [D-Arg0, Hyp3, Thi5,8, D-Phe7]-Adamantaneacetyl Bradykinin
Molecular Weight 1469.75
Storage Requirements -30° to -10°C
Sequence N-Adamantaneacetyl-D-Arg-Arg-Pro-Hyp-Gly-Thi-Ser-D-Phe-Thi-Arg
Format Solid
Common Name [N-Admantaneacetyl-D-Arg0-Hyp3, Thi58, D-Phe7] Bradykinin
Regulatory Status RUO
Purification Method HPLC