Please login to your online account to display your discounted pricing

Alfa Aesar™ [D-Arg&superscript_1;, D-Phe&superscript_5;, D-Trp&superscript_7;,&superscript_9;, Leu&superscript_1;&superscript_1;] Substance P

For cellular and molecular biology applications

Specifications

Product Type [D-Arg1, D-Phe5, D-Trp7,9, Leu11] Substance P
Molecular Weight 1516.85
Storage Requirements -30° to -10°C
Format Solid
Sequence D-Arg-Pro-Lys-Pro-D-Phe-Gln-D-Trp-Phe-D-Trp-Leu-Leu-NH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66988LB0

 
alfa aesar™
J66988LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66988MCR

 
alfa aesar™
J66988MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [D-Arg1, D-Phe5, D-Trp7,9, Leu11] Substance P
Molecular Weight 1516.85
Storage Requirements -30° to -10°C
Format Solid
Sequence D-Arg-Pro-Lys-Pro-D-Phe-Gln-D-Trp-Phe-D-Trp-Leu-Leu-NH2
Common Name Antagonist D
Regulatory Status RUO
Purification Method HPLC