Please login to your online account to display your discounted pricing

Alfa Aesar™ [D-Arg&superscript_1;, D-Phe&superscript_5;, D-Trp&superscript_7;,&superscript_9;, Leu&superscript_1;&superscript_1;] Substance P


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66988LB0

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66988LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 5mg
Format Solid
Sequence D-Arg-Pro-Lys-Pro-D-Phe-Gln-D-Trp-Phe-D-Trp-Leu-Leu-NH2
Regulatory Status RUO
Storage Requirements -30° to -10°C
Common Name Antagonist D
Product Type [D-Arg1, D-Phe5, D-Trp7,9, Leu11] Substance P
Molecular Weight 1516.85
Purification Method HPLC