Please login to your online account to display your discounted pricing

D-(-)-Arabinose (White Powder), Fisher BioReagents


Quantity 25g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP250425

 View more versions of this product

Catalog No. BP250425

  • / Each
  • / Case of 8 Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder/Solid
Formula Weight 150.07
Melting Point 152°C
Quantity 25g
Molecular Formula C5H10O5
Color White
Identification Pass Test
Chemical Name or Material D-(-)-Arabinose
FTIR Conforms to reference
Assay Percent Range ≥99 %
Packaging Amber Glass
CAS 28697-53-2
ChemAlert Storage Symbol Gray
Purity 99% min.
Specific Rotation -107 to -97° (+ or -)
Recommended Storage RT

Specifications

Chemical Name or Material D-(-)-Arabinose
CAS 28697-53-2
Physical Form Powder/Solid
Assay Percent Range ≥99 %
Molecular Formula C5H10O5
Formula Weight 150.07
Accessories
 

RT

SDS & Safety Documents