missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-(-)-Arabinose (White Powder), Fisher BioReagents

$1,008.37 - $1,008.37

Chemical Identifiers

CAS 28697-53-2
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00064361,MFCD20731172,MFCD00006609,MFCD00066648,MFCD09864633
InChI Key PYMYPHUHKUWMLA-DEOSMSJNSA-N
Synonym 2s,3s,4r,5r-oxane-2,3,4,5-tetrol, 2s,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol, a-d-arabinopyranose, alpha-arabinose, alpha-d-arabinopyranose, alpha-d-arabinopyranose 9ci, d--arabinose
PubChem CID 71313478
IUPAC Name (4S,5R)-oxane-2,3,4,5-tetrol
SMILES OCC(O)[C@@H](O)[C@H](O)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP2504100
SDS View Documents
Fisher BioReagents
BP2504100
100g Amber Glass
Each for $1,008.37
Case of 3 Each for $3,025.11
 
Description

Description

D-(-)-Arabinose is a component used for culture media.
Specifications

Chemical Identifiers

28697-53-2
150.13
PYMYPHUHKUWMLA-DEOSMSJNSA-N
71313478
OCC(O)[C@@H](O)[C@H](O)C=O
C5H10O5
MFCD00064361,MFCD20731172,MFCD00006609,MFCD00066648,MFCD09864633
2s,3s,4r,5r-oxane-2,3,4,5-tetrol, 2s,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol, a-d-arabinopyranose, alpha-arabinose, alpha-d-arabinopyranose, alpha-d-arabinopyranose 9ci, d--arabinose
(4S,5R)-oxane-2,3,4,5-tetrol

Specifications

Pass Test
-107 to -97° (+ or -)
152°C
28697-53-2
C5H10O5
2s,3s,4r,5r-oxane-2,3,4,5-tetrol, 2s,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol, a-d-arabinopyranose, alpha-arabinose, alpha-d-arabinopyranose, alpha-d-arabinopyranose 9ci, d--arabinose
OCC(O)[C@@H](O)[C@H](O)C=O
150.13
150.07
99% min.
Amber Glass
White
100g
≥99 %
MFCD00064361,MFCD20731172,MFCD00006609,MFCD00066648,MFCD09864633
PYMYPHUHKUWMLA-DEOSMSJNSA-N
(4S,5R)-oxane-2,3,4,5-tetrol
71313478
Powder/Solid
D-(-)-Arabinose
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS
Product Certifications