Please login to your online account to display your discounted pricing

D-AP7, Tocris


Specific NMDA antagonist. More active form of DL-AP7

Quantity 10mg

Manufacturer: r & d systems  RDS016410

 View more versions of this product

Catalog No. 016410

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 225.18
Assay Percent Range >98%
Chemical Name or Material D-AP7
Synonym D-(-)-2-Amino-7-phosphonoheptanoic acid
CAS 81338-23-0
Molecular Formula C7H16NO5P
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material D-AP7
Assay Percent Range >98%
Molecular Formula C7H16NO5P
Formula Weight 225.18
Synonym D-(-)-2-Amino-7-phosphonoheptanoic acid
Accessories

Store at Room Temperature