missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-AP7, Tocris

Specific NMDA antagonist. More active form of DL-AP7

$306.33 - $306.33

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C7H16NO5P
Formula Weight 225.18
Synonym D-(-)-2-Amino-7-phosphonoheptanoic acid
Chemical Name or Material D-AP7
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
016410 R & D Systems
RDS016410
10mg Each for $306.33
Specifications Description & Specifications

Chemical Identifiers

>98%
225.18
D-AP7
C7H16NO5P
D-(-)-2-Amino-7-phosphonoheptanoic acid

Specifications

81338-23-0
>98%
225.18
D-AP7
10mg
C7H16NO5P
D-(-)-2-Amino-7-phosphonoheptanoic acid