Please login to your online account to display your discounted pricing

D-AP5, Tocris

Potent, selective NMDA antagonist; More active form of DL-AP5

$93.40 - $1,112.69

Chemical Identifiers

Chemical Name or Material D-AP5
Assay Percent Range >99%
Molecular Formula C5H12NO5P
Formula Weight 197.13
Synonym D-(-)-2-Amino-5-phosphonopentanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

01061

  SDS
r & d systems
RDS01061
1mg Each for $93.40

010610

 
r & d systems
RDS010610
10mg Each for $195.49

0106100

 
r & d systems
RDS0106100
100mg Each for $1,112.69

010650

 
r & d systems
RDS010650
50mg Each for $816.90
Description & Specifications

Specifications

Quantity 1mg
Formula Weight 197.13
Assay Percent Range >99%
Chemical Name or Material D-AP5
Molecular Formula C5H12NO5P
CAS 79055-68-8
Synonym D-(-)-2-Amino-5-phosphonopentanoic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature