Learn More
D-Amygdalin hydrate, 96%, Thermo Scientific Chemicals
$54.36 - $132.21
Chemical Identifiers
CAS | 29883-15-6 |
---|---|
Molecular Formula | C20H27NO11 |
Molecular Weight (g/mol) | 457.43 |
MDL Number | MFCD29049822 |
InChI Key | XUCIJNAGGSZNQT-ARGBPYSKNA-N |
Synonym | amygdalin |
PubChem CID | 66509032 |
IUPAC Name | 2-phenyl-2-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxan-2-yl]oxy}acetonitrile |
SMILES | OC[C@H]1O[C@@H](OC[C@H]2O[C@@H](OC(C#N)C3=CC=CC=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1273104
|
Thermo Scientific Chemicals
L1273104 |
2 g |
Each for $54.36
|
|
|||||
AAL1273109
|
Thermo Scientific Chemicals
L1273109 |
10 g |
Each for $132.21
|
|
|||||
Description
A benzylic glycoside that has been used as an antineoplastic agent
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
29883-15-6 | |
457.43 | |
XUCIJNAGGSZNQT-ARGBPYSKNA-N | |
66509032 | |
OC[C@H]1O[C@@H](OC[C@H]2O[C@@H](OC(C#N)C3=CC=CC=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O |
C20H27NO11 | |
MFCD29049822 | |
amygdalin | |
2-phenyl-2-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxan-2-yl]oxy}acetonitrile |
Specifications
29883-15-6 | |
C20H27NO11 | |
2 g | |
14,597 | |
XUCIJNAGGSZNQT-ARGBPYSKNA-N | |
OC[C@H]1O[C@@H](OC[C@H]2O[C@@H](OC(C#N)C3=CC=CC=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O | |
457.43 | |
457.44 (Anhydrous) | |
D-Amygdalin hydrate |
215°C to 220°C | |
MFCD29049822 | |
66856 | |
amygdalin | |
−36° (c=1.2 in Water) | |
2-phenyl-2-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxan-2-yl]oxy}acetonitrile | |
66509032 | |
96% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 249-925-3
RTECSNumber : OO8450000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only