missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-allo-Threonine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H27050MD
Description
They are found as constituents of an increasing group of biologically active peptides. Stereocontrolled syntheses of chiral and racemic key intermediates to thienamycin from d-allo-threonine and trans-crotonic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThey are found as constituents of an increasing group of biologically active peptides. Stereocontrolled syntheses of chiral and racemic key intermediates to thienamycin from d-allo-threonine and trans-crotonic acid.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
268°C (decomposition) | |
24830-94-2 | |
MFCD00004526 | |
d-allo-threonine, d-allothreonine, h-d-allo-thr-oh, d--allo-threonine, r-allothreonine, 2r,3r-2-amino-3-hydroxybutanoic acid, 2r,3r-allothreonine, r-allo-threonine, dl-allo-threonine, allothreonine, d | |
AYFVYJQAPQTCCC-STHAYSLISA-N | |
(2R,3R)-2-amino-3-hydroxybutanoic acid | |
90624 | |
119.12 | |
250 mg |
D-allo-Threonine | |
C4H9NO3 | |
1721644 | |
Soluble in water. | |
C[C@H](O)[C@@H](N)C(O)=O | |
119.12 | |
CHEBI:32826 | |
99% | |
−8.7° (c=2 in Water) |
Chemical Identifiers
24830-94-2 | |
119.12 | |
AYFVYJQAPQTCCC-STHAYSLISA-N | |
90624 | |
(2R,3R)-2-amino-3-hydroxybutanoic acid |
C4H9NO3 | |
MFCD00004526 | |
d-allo-threonine, d-allothreonine, h-d-allo-thr-oh, d--allo-threonine, r-allothreonine, 2r,3r-2-amino-3-hydroxybutanoic acid, 2r,3r-allothreonine, r-allo-threonine, dl-allo-threonine, allothreonine, d | |
CHEBI:32826 | |
C[C@H](O)[C@@H](N)C(O)=O |
Safety and Handling
EINECSNumber : 246-488-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only