missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Alanine tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals
$160.77 - $502.81
Chemical Identifiers
CAS | 59531-86-1 |
---|---|
Molecular Formula | C7H16ClNO2 |
Molecular Weight (g/mol) | 181.66 |
MDL Number | MFCD00077087 |
InChI Key | WIQIWPPQGWGVHD-NUBCRITNSA-N |
Synonym | d-alanine tert-butyl ester hydrochloride, h-d-ala-otbu.hcl, r-tert-butyl 2-aminopropanoate hydrochloride, r-2-aminopropionic acid tert-butyl ester hydrochloride, tert-butyl 2r-2-aminopropanoate hydrochloride, r-alanine t-butyl ester hcl, d-alanine tert-butyl ester hcl, d-alanine t-butyl ester hydrochloride, d-alanine-otbu hcl, d-alanine, 1,1-dimethylethyl ester, hydrochloride |
PubChem CID | 16211919 |
IUPAC Name | tert-butyl (2R)-2-aminopropanoate;hydrochloride |
SMILES | CC(C(=O)OC(C)(C)C)N.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5989803
|
Thermo Scientific Chemicals
H5989803 |
1 g |
Each for $160.77
|
|
|||||
AAH5989806
|
Thermo Scientific Chemicals
H5989806 |
5 g |
Each for $502.81
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
59531-86-1 | |
181.66 | |
WIQIWPPQGWGVHD-NUBCRITNSA-N | |
16211919 | |
CC(C(=O)OC(C)(C)C)N.Cl |
C7H16ClNO2 | |
MFCD00077087 | |
d-alanine tert-butyl ester hydrochloride, h-d-ala-otbu.hcl, r-tert-butyl 2-aminopropanoate hydrochloride, r-2-aminopropionic acid tert-butyl ester hydrochloride, tert-butyl 2r-2-aminopropanoate hydrochloride, r-alanine t-butyl ester hcl, d-alanine tert-butyl ester hcl, d-alanine t-butyl ester hydrochloride, d-alanine-otbu hcl, d-alanine, 1,1-dimethylethyl ester, hydrochloride | |
tert-butyl (2R)-2-aminopropanoate;hydrochloride |
Specifications
59531-86-1 | |
MFCD00077087 | |
d-alanine tert-butyl ester hydrochloride, h-d-ala-otbu.hcl, r-tert-butyl 2-aminopropanoate hydrochloride, r-2-aminopropionic acid tert-butyl ester hydrochloride, tert-butyl 2r-2-aminopropanoate hydrochloride, r-alanine t-butyl ester hcl, d-alanine tert-butyl ester hcl, d-alanine t-butyl ester hydrochloride, d-alanine-otbu hcl, d-alanine, 1,1-dimethylethyl ester, hydrochloride | |
CC(C(=O)OC(C)(C)C)N.Cl | |
181.66 | |
181.66 | |
1 g | |
Hygroscopic |
C7H16ClNO2 | |
5763077 | |
WIQIWPPQGWGVHD-NUBCRITNSA-N | |
tert-butyl (2R)-2-aminopropanoate;hydrochloride | |
16211919 | |
98% | |
−1.4° (c=2 in Ethanol) | |
D-Alanine tert-butyl ester hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only