missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Alanine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6247214
Description
D-Alanine methyl ester hydrochloride is used as building block for the preparation of peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Alanine methyl ester hydrochloride is used as building block for the preparation of peptides.
Solubility
Soluble in Water 100 mg/ml.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
108°C to 110°C | |
14316-06-4 | |
C4H10ClNO2 | |
4235975 | |
Soluble in Water 100mg/ml. | |
[H+].[Cl-].COC(=O)C(C)N | |
139.58 | |
139.58 | |
25 g |
D-Alanine methyl ester hydrochloride | |
White | |
MFCD00066141 | |
d-alanine methyl ester hydrochloride, methyl d-alaninate hydrochloride, h-d-ala-ome.hcl, methyl d-alaninate hcl, methyl 2r-2-aminopropanoate hydrochloride, h-d-ala-ome hcl, r-methyl 2-aminopropanoate hydrochloride, d-alanine methylester hydrochloride, d-alanine, methyl ester, hydrochloride, pubchem10885 | |
IYUKFAFDFHZKPI-UHFFFAOYNA-N | |
methyl (2R)-2-aminopropanoate;hydrochloride | |
11182647 | |
98% | |
Crystalline Powder |
Chemical Identifiers
14316-06-4 | |
139.58 | |
IYUKFAFDFHZKPI-UHFFFAOYNA-N | |
11182647 | |
[H+].[Cl-].COC(=O)C(C)N |
C4H10ClNO2 | |
MFCD00066141 | |
d-alanine methyl ester hydrochloride, methyl d-alaninate hydrochloride, h-d-ala-ome.hcl, methyl d-alaninate hcl, methyl 2r-2-aminopropanoate hydrochloride, h-d-ala-ome hcl, r-methyl 2-aminopropanoate hydrochloride, d-alanine methylester hydrochloride, d-alanine, methyl ester, hydrochloride, pubchem10885 | |
methyl (2R)-2-aminopropanoate;hydrochloride |
Safety and Handling
EINECSNumber : 238-258-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only